Learn More
Lenz Laborglasintrumente Round Bottom Flasks
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07035094
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11983621
  • € 470.64 / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN ISO 24450 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Specifications
89 mm
20 L
Duran Glass
520 mm