Learn More
Witeg™ Round Bottom Flasks
Product Code. 10296753
Change view
Click to view available options
Capacity: 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10296753 Capacity: 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10296753 Supplier Witeg™ Supplier No. 0 603 100 S
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
With a center neck for socket ST 29/32.
  • 64 x 110 mm
  • Acc. to DIN 12348
  • Provided in packs of 10
Specifications
Certifications/Compliance DIN 12348
Color Transparent
Capacity (Metric) 100 mL
Material Glass
No. per Pack 10
Product Type Flask
Socket Joint ST 29/32
Bottom Shape Round