Learn More
Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation
Product Code. 10767571
Click to view available options
Closure Type:
Not Included
Stopper
Unit Size:
Pack of 10
Product Code. 10767571
Brand: Lenz Laborglasinstrumente™ 05700107
Ideal for routine laboratory analysis and procedures. Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation are manufactured from durable DURAN™ borosilicate glass.
  • With round bottom and NS socket
  • Available without stopper, with PE stopper, or with hexagonal hollow glass stopper
  • Corrosion-free and resistant to thermal shock
Specifications
Diameter (Metric) 16 mm
Product Type Glass Tube
Material Borosilicate Glass
Closure Material HDPE
Height (Metric) 100 mm
Closure Type Stopper