Learn More
Buerkle™ Round Containers
Product Code. p-200115231 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
14 L
100 L
250 L
300 L
Unit Size:
Each
Product Code. 17908451
Brand: Buerkle™ 34900144
Only null left
Add to basket
Add to basket
Container for various applications. Produced seamlessly and stress-relieved with the sintering method. The wall thickness increases downwardly. This guarantees optimum ratio of wall thickness to filling load. Custom-made, exchange excluded.
  • L-LDPE (linear LDPE), transparent
  • Rupture-proof
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer 24.5 cm
Type Tank
Material LLDPE
Capacity (Metric) 14 L
Height (Metric) 32.5 cm