Learn More
Round Glass Narrow Mouth Alpha Bottles, without Cap
Product Code. p-8015547
Click to view available options
Capacity (Metric):
30 mL
125 mL
Color:
Amber
Clear
Unit Size:
Pack of 104
Pack of 156
Product Code. 12800914
Brand: SGD PHARMA 300776
Store, pour or share your laboratory liquids using Round Glass Narrow Mouth Alpha Bottles, without Cap. Made from amber or clear borosilicate glass, the round bottles are an integral tool in the laboratory.
  • Narrow neck
  • Borosilicate glass
  • Round
Specifications
Capacity (Metric) 30 mL
Color Amber
Material Borosilicate Glass
No. per Pack 40Pack
Quantity 40Pack
Mouth Narrow
Height (Metric) 67 mm
Product Type Alpha Bottle
Closure Liner Polypropylene/SOP
Closure Material Polypropylene
Diameter (Metric) 35 mm