Learn More
Emma Safety Footwear Roy All-Round Protection Versatile D-Fit Shoes
Designed with wipe-clean uppers and easy-clean sole.
Brand:  Emma Safety Footwear 700540/38
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13254504
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Full grain leather uppers
  • Hydro-Tec™ sanitized silver lining
  • Non-woven antistatic insole
  • GripForce™ Easy Twist outsole
Specifications
Specifications
Hydro-Tec Sanitized Silver lining
Non-woven antistatic insole
Steel Toe
38
GripForce™ Easy Twist outsole
All-Round Protection Versatile Shoes