missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RP105/CD180 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76706AF532
This item is not returnable.
View return policy
Description
RP105/CD180 Polyclonal antibody specifically detects RP105/CD180 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RP105/CD180 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| CD180 antigenMGC126233, CD180 molecule, LY64MGC126234, Ly78, Lymphocyte antigen 64, lymphocyte antigen-64, radioprotective, 105kDa, Radioprotective 105 kDa protein, RP105lymphocyte antigen 64 (mouse) homolog, radioprotective, 105kD | |
| Antibody was raised against a 11 amino acid synthetic peptide from near the amino terminus of human RP105. The immunogen is located within the first 50 amino acids of RP105. Amino Acid Squence: CIEKEANKTYN | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Innate Immunity | |
| 4064 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction