missing translation for 'onlineSavingsMsg'
Learn More

RP105/CD180 Antibody (G28-8) - Low Endotoxin - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-62228

Product Code. 18647346

  • ONLINE EXCLUSIVE PRICE
    € 256.00
    € 241.50 / 0.10mg
    Save € 14.50
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RP105/CD180 Monoclonal antibody specifically detects RP105/CD180 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Functional Assay
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RP105/CD180
Monoclonal
1 mg/mL
Flow Cytometry 2 μg/ml, Immunoprecipitation 1:10-1:500, Functional
CD180 antigenMGC126233, CD180 molecule, LY64MGC126234, Ly78, Lymphocyte antigen 64, lymphocyte antigen-64, radioprotective, 105kDa, Radioprotective 105 kDa protein, RP105lymphocyte antigen 64 (mouse) homolog, radioprotective, 105kD
Human tonsillar B cells
0.1 mg
Adaptive Immunity, Immunology, Innate Immunity
4064
Store at 4°C. Do not freeze.
IgG1 κ
Flow Cytometry, Immunoprecipitation, Functional Assay
G28-8
Unconjugated
PBS (pH 7.4), 0.2 um filter sterilized
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.