Learn More
Invitrogen™ RP2 Monoclonal Antibody (3D7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA536239
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 tissue, human Jurkat, whole cell, human Caco-2 whole cell, human Hela whole cell, human A549 whole cell. ICC/IF: U2OS cell. Flow: 293T cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The RP2 locus has been implicated as one cause of X-linked retinitis pigmentosa. The predicted gene product shows homology with human cofactor C, a protein involved in the ultimate step of beta-tubulin folding.
Specifications
| RP2 | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| O75695, Q9EPK2 | |
| RP2 | |
| E. coli-derived human RP2recombinant protein (Position: D244-M348). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 3D7 | |
| Unconjugated | |
| RP2 | |
| AI662636; DELXp11.3; KIAA0215; NM23-H10; NME10; Protein XRP2; retinitis pigmentosa 2 (X-linked recessive); retinitis pigmentosa 2 homolog (human); RGD1565124; Rp2; RP2 activator of ARL3 GTPase; RP2, ARL3 GTPase activating protein; Rp2h; TBCCD2; XRP2; XRP2 protein | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 19889, 367714, 6102 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.