missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPA14 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97782CL1
This item is not returnable.
View return policy
Description
RPA14 Polyclonal antibody specifically detects RPA14 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| RPA14 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| REPA3Replication factor A protein 3, replication protein A 14 kDa subunit, replication protein A3 (14kD), replication protein A3, 14kDa, RF-A protein 3, RP-A p14, RPA14 | |
| Produced in rabbits immunized with E. coli-derived Human RPA14 fragment. | |
| 0.1 mL | |
| Cancer, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers | |
| 6119 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction