missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPA14 Monoclonal Antibody (14.2)
GREENER_CHOICE

Product Code. 11564692
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11564692 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11564692 Supplier Invitrogen™ Supplier No. MA123281

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Recommended positive controls: MCF7, T24.

Replication protein A (RPA) is a single-stranded DNA binding protein. Human RPA is a heterotrimeric protein containing subunits of 70, 32 and 14 kDa. This protein complex is highly conserved in eukaryotes and is essential in DNA replication, homologous recombination and nucleotide excision repair. RPA has been shown to be a target for DNA-dependent protein kinase (DNA-PK) ataxia telangiectasia-mutated gene (ATM) and a cyclin dependent protein kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPA14
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 14.2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Rpa3
Gene Accession No. P35244, Q9CQ71
Gene Alias 14kDa; C330026P08Rik; REPA3; Replication factor A protein 3; replication protein A 14 kDa subunit; replication protein A3; replication protein A3, 14kDa; RF-A protein 3; RP-A p14; RPA14; RPA3
Gene Symbols Rpa3
Host Species Mouse
Immunogen Full-length human RPA-14 expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6119, 68240
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.