missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RPA2 Monoclonal specifically detects RPA2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | RPA2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | MA34 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Immunohistochemistry 1:10 - 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin 1:10 - 1:100 |
| Formulation | PBS with 1mg/ml BSA with 0.05% Sodium Azide |
| Gene Alias | REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34 |
| Gene Symbols | RPA2 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?