missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPA2 Antibody (RPA2/3140R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08330AF700
This item is not returnable.
View return policy
Description
RPA2 Monoclonal antibody specifically detects RPA2 in Human samples. It is validated for Western Blot
Specifications
| RPA2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34 | |
| Recombinant human full-length Replication protein A2 (RPA2) protein (Uniprot: P15927) | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers | |
| 6118 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| RPA2/3140R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction