missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPA70 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35085-100ul
This item is not returnable.
View return policy
Description
RPA70 Polyclonal antibody specifically detects RPA70 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| RPA70 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:100 | |
| HSSB, MST075, MSTP075, REPA1RF-A protein 1, Replication factor A protein 1, replication protein A 70 kDa DNA-binding subunit, replication protein A1 (70kD), replication protein A1, 70kDa, RF-A, RP-A, RPA70RP-A p70, Single-stranded DNA-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human RPA70 (NP_002936.1).,, Sequence:, MVGQLSEGAIAAIMQKGDTNIKPILQVINIRPITTGNSPPRYRLLMSDGLNTLSSFMLATQLNPLVEEEQLSSNCVCQIHRFIVNTLKDGRRVVILMELEVLKSAEAVGVKIGNPVPYNEGLGQPQVAPPAPAASPAASSRPQPQNGSSGMGSTVSKAYGASKTFGKAAGPSLSHTSGGTQSKVVPIASLTPYQSKWTICARVTNKSQIRTWSNSRGEGKLFSLELVDESGEIRATAFNE | |
| 100 μL | |
| DNA Repair, Nucleotide Excision Repair | |
| 6117 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction