missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ rpaA Polyclonal Antibody
GREENER_CHOICE

Product Code. 17979292
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17979292 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17979292 Supplier Invitrogen™ Supplier No. PA5144443

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

rpaA Polyclonal Antibody for Western Blot, ELISA

RpaA is a gene that encodes a transcriptional regulator protein in bacteria. It is involved in the regulation of several cellular processes, including stress response, metabolism, and quorum sensing. The precise function of RpaA can vary depending on the bacterial species and the environmental conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen rpaA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias rpaA; Two-component response regulator
Host Species Rabbit
Immunogen Recombinant Nostoc sp. (strain PCC 7120/SAG 25.82/UTEX 2576) Two-component response regulator (1-247aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.