Learn More
RPAIN Mouse, anti-Human, Clone: 2D8E3, Proteintech
Product Code. 18015155 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015155 20 μL 20µL
18097265 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18015155 Supplier Proteintech Supplier No. 685461IG20UL
Mouse Monoclonal Antibody
The RPAIN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPAIN. This antibody recognizes human antigen. The RPAIN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Antigen RPAIN
Applications Western Blot, ELISA
Clone 2D8E3
Concentration 1000 μg/mL
Gene Symbols HRIP, RIP, RPA interacting protein, RPAIN
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 84268
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG1