missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPC62 Antibody (OTI2H1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73942AF594
This item is not returnable.
View return policy
Description
RPC62 Monoclonal specifically detects RPC62 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| RPC62 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| POLR3C | |
| Full length recombinant protein of human RPC62 (NP_006459) produced in HEK293T cells. | |
| 0.1 mL | |
| 10623.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2H1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DNA-directed III 62 kDa polypeptide, DNA-directed RNA polymerase III subunit C, DNA-directed RNA polymerase III subunit RPC3, polymerase (RNA) III (DNA directed) polypeptide C (62kD), RNA polymerase III 62 kDa subunit, RNA polymerase III subunit C3, RPC62RPC3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction