missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPE65 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95096-0.1ml
This item is not returnable.
View return policy
Description
RPE65 Polyclonal antibody specifically detects RPE65 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| RPE65 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| All-trans-retinyl-palmitate hydrolase, BCO family, member 3, BCO3, EC 3.1.1.64, EC:3.1.1.64, EC:5.3.3.22, LCA2, lutein isomerase, meso-zeaxanthin isomerase, mRPE65, p63, RBP-binding membrane protein, rd12, retinal pigment epithelium specific protein 65, Retinal pigment epithelium-specific 65 kDa protein, retinal pigment epithelium-specific protein 65kDa, retinitis pigmentosa 20 (autosomal recessive), retinoid isomerohydrolase, Retinol isomerase, RP20, RPE65, RPE65, retinoid isomerohydrolase, sRPE65 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 274-533 of human RPE65 (NP_000320.1). NYMDCFESNETMGVWLHIADKKRKKYLNNKYRTSPFNLFHHINTYEDNGFLIVDLCCWKGFEFVYNYLYLANLRENWEEVKKNARKAPQPEVRRYVLPLNIDKADTGKNLVTLPNTTATAILCSDETIWLEPEVLFSGPRQAFEFPQINYQKYCGKPYTYAYGLGLNHFVPDRLCKLNVKTKETWVWQEPDSYPSEPIFVSHPDALEEDDGVVLSVVVSPGAGQKPAYLLILNAKDLSEVARAEVEINIPVTFHGLFKKS | |
| 0.1 mL | |
| Cellular Markers, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Sensory Systems, Signal Transduction, Vision | |
| 6121 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction