Learn More
Invitrogen™ RPE65 Monoclonal Antibody (401.8B11.3D9), DyLight™ 650
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA516041
Product Code. 13217658
  • € 582.00 / 200µL
Description
RPE65 Monoclonal Antibody for Western Blot, IHC (P), IHC (F), ICC/IF, Flow, IP
RPE65 is a major protein of the retinal pigment epithelium (RPE). RPE65 is essential for the regeneration of rhodopsin in the visual cycle. Mutations in RPE65 are responsible for certain forms of autosomal recessive severe retinal dystrophy, including Leber Congenital Amaurosis (LCA). Loss of function results in a block in retinoid processing required for the synthesis of 11-cis retinal and the accumulation of retinyl esters in the RPE.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPE65
Monoclonal
0.64 mg/mL
50mM sodium borate with 0.05% sodium azide
O70276, Q16518, Q28175, Q91ZQ5, Q9TVB8, Q9YGX2
RPE65
Bovine RPE65 microsomal membrane proteins.
200 μL
Primary
Human, Mouse, Rat, Canine, Monkey, Bovine, Pig, Chicken, Frog
Antibody
IgG1 κ
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
401.8B11.3D9
DyLight 650
RPE65
65kDa; A930029L06Rik; all-trans-retinyl-palmitate hydrolase; BCO family, member 3; BCO3; LCA2; Lutein isomerase; membrane receptor p63; meso-zeaxanthin isomerase; modifier of retinal degeneration 1; Mord1; mRPE65; p63; RBP-binding membrane protein; rd12; retinal pigment epithelium 65; retinal pigment epithelium 65-like; retinal pigment epithelium abundant protein RPE65; retinal pigment epithelium specific protein 65; retinal pigment epithelium, 65 kDa; retinal pigment epithelium-specific 65 kDa protein; retinal pigment epithelium-specific 65kD protein; retinal pigment epithelium-specific protein (65kD); retinal pigment epithelium-specific protein 65kDa; retinal pigment epithelium-specific protein RPE65; Retinitis pigmentos; retinitis pigmentosa 20 (autosomal recessive); retinoid isomerohydrolase; retinoid isomerohydrolase RPE65; retinol isomerase; RP20; RPE65; rpe65 gene for retinal pigment epithelium-specific protein; RPE65, retinoid isomerohydrolase; sRPE65; truncated RPE65
Mouse
Protein G
RUO
100516743, 19892, 282043, 395700, 403803, 6121, 89826
4°C, store in dark