Learn More
RPE65 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-17939-20UL
 View more versions of this product
Product Code. 18028415
  • € 145.00 / 20µL
Description
The RPE65 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RPE65. This antibody recognizes human, mouse, rat antigen. The RPE65 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
RPE65
1000 μg/mL
LCA2, mRPE65, rd12, Retinoid isomerohydrolase, Retinol isomerase, RP20, RPE65, sRPE65
20 μL
6121
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa