missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPIA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598374
This item is not returnable.
View return policy
Description
RPIA Polyclonal Antibody for Western Blot, ELISA
Defects in RPIA are the cause of ribose 5-phosphate isomerase deficiency. The exact function of RPIA remains unknown.
Specifications
| RPIA | |
| Polyclonal | |
| Unconjugated | |
| Phosphoriboisomerase; ribose 5-phosphate epimerase; ribose 5-phosphate isomerase A; ribose 5-phosphate isomerase A (ribose 5-phosphate epimerase); ribose-5-phosphate isomerase; RPI; Rpia; RPIAD | |
| Recombinant Escherichia coli Ribose-5-phosphate isomerase A protein (1-219AA). | |
| 100 μg | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction