missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPIA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112699
This item is not returnable.
View return policy
Description
RPIA Polyclonal Antibody for Western Blot, IHC (P), ELISA
Defects in RPIA are the cause of ribose 5-phosphate isomerase deficiency. The exact function of RPIA remains unknown.
Specifications
| RPIA | |
| Polyclonal | |
| Unconjugated | |
| RPIA | |
| Phosphoriboisomerase; ribose 5-phosphate epimerase; ribose 5-phosphate isomerase A; ribose 5-phosphate isomerase A (ribose 5-phosphate epimerase); ribose-5-phosphate isomerase; RPI; Rpia; RPIAD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19895, 22934 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.05 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P47968, P49247 | |
| RPIA | |
| Recombinant Human Ribose-5-phosphate isomerase protein (82-311AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction