missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPIP8 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97453FR
This item is not returnable.
View return policy
Description
RPIP8 Polyclonal specifically detects RPIP8 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| RPIP8 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| Rap2-interacting protein 8, RAP2IPRUN domain-containing protein 3A, RPIP-8, RPIP8RaP2 interacting protein 8, RUN domain containing 3A | |
| Produced in rabbits immunized with E. coli-derived Human RPIP8 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 10900 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction