missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Antibody (LAMP3/8770R), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21028JF646
This item is not returnable.
View return policy
Description
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD63 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
| Recombinant full-length human CD63 protein | |
| 0.1 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
| 967 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| LAMP3/8770R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction