missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPIP8 Antibody (OTI3C6), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73943AF647
This item is not returnable.
View return policy
Description
RPIP8 Monoclonal specifically detects RPIP8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RPIP8 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RUNDC3A | |
| Full length recombinant protein of human RPIP8 (NP_006686) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3C6 | |
| Western Blot, Immunohistochemistry | |
| Rap2-interacting protein 8, RAP2IPRUN domain-containing protein 3A, RPIP-8, RPIP8RaP2 interacting protein 8, RUN domain containing 3A | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 10900.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction