Learn More
RPL10 Mouse anti-Human, Clone: OTI9A8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807673
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a ribosomal protein that is a component of the 60S ribosome subunit. The related protein in chicken can bind to c-Jun and can repress c-Jun-mediated transcriptional activation. Some studies have detected an association between variation in this gene and autism spectrum disorders, though others do not detect this relationship. There are multiple pseudogenes of this gene dispersed throughout the genome. Alternative splicing results in multiple transcript variants.Specifications
| RPL10 | |
| Monoclonal | |
| Unconjugated | |
| RPL10 | |
| AUTSX5, DXS648, DXS648E, L10, MRXS35, NOV, QM | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6134 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9A8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P27635 | |
| RPL10 | |
| Full length human recombit protein of human RPL10 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.