missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPL13A Polyclonal Antibody
GREENER_CHOICE

Product Code. 15786401 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15786401 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15786401 Supplier Invitrogen™ Supplier No. PA558528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AHEVGWKYQA VTATLEEKRK EKAKIHYRKK KQLMRLRKQA EKNV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 98%.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L13P family of ribosomal proteins. It is located in the cytoplasm. Transcript variants utilizing alternative polyA signals have been observed. This gene is co-transcribed with the small nucleolar RNA genes U32, U33, U34, and U35, which are located in its second, fourth, fifth, and sixth introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL13A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL13A
Gene Accession No. P40429
Gene Alias 1810026N22Rik; 23 kDa highly basic protein; 60S ribosomal protein L13a; L13A; Large ribosomal subunit protein uL13; P198; ribosomal protein L13a; Rpl13a; tissue specific transplantation antigen 1; tissue specific transplantation antigen P198-7; transplantation antigen P198; TSTA1; Tstap198-7; tum-antigen; Tum-P198 antigen; tum-transplantation antigen P198
Gene Symbols RPL13A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL13A. Recombinant protein control fragment (Product #RP-98826).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23521
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.