missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552058
This item is not returnable.
View return policy
Description
Immunogen sequence: KKMRNRIIKN EVKKLQKAAL LKASPKKAPG TKGTAAAAAA AAAAAAAAAK VPAKKITAAS KKAPAQKVPA QKATGQKAAP APKAQKGQKA PAQKAPAPKA S Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L14E family of ribosomal proteins. It contains a basic region-leucine zipper (bZIP)-like domain. The protein is located in the cytoplasm. This gene contains a trinucleotide (GCT) repeat tract whose length is highly polymorphic; these triplet repeats result in a stretch of alanine residues in the encoded protein. Transcript variants utilizing alternative polyA signals and alternative 5'-terminal exons exist but all encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Aoki M., Diabetes 45:157-164(1996). Tanaka M., Biochem. Biophys. Res. Commun. 243:531-537(1998). Yoshihama M., Genome Res. 12:379-390(2002).
Specifications
| RPL14 | |
| Polyclonal | |
| Unconjugated | |
| RPL14 | |
| 3100001N19Rik; 60S ribosomal protein L14; AA407502; AL022816; CAG-ISL 7; CAG-ISL-7; CTG-B33; hRL14; L14; Large ribosomal subunit protein eL14; MGC88594; ribosomal protein L14; RL14; RPL14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9045 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P50914 | |
| RPL14 | |
| Recombinant protein corresponding to Human RPL14. Recombinant protein control fragment (Product #RP-102221). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction