missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPL17 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16320635
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16320635 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16320635 Supplier Invitrogen™ Supplier No. PA583677

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KNTRETAQAI KGMHIRKATK YLKDVTLQKQ CVPFRRYNGG VGRCAQAKQW GWTQGRWPKK SAEFLLHMLK NAESNAELKG LDVDSLVIEH IQVNKAPKM Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L22P family of ribosomal proteins. It is located in the cytoplasm. This gene has been referred to as rpL23 because the encoded protein shares amino acid identity with ribosomal protein L23 from Halobacterium marismortui; however, its official symbol is RPL17. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternative splicing results in multiple transcript variants. Read-through transcription also exists between this gene and the neighboring downstream C18orf32 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL17
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rpl17
Gene Accession No. P18621
Gene Alias 60S ribosomal protein L17; 60S ribosomal protein L23; Amino acid starvation-induced protein; ASI; gene encoding putative NFkB activating protein; L17; L23; Large ribosomal subunit protein uL22; MGC117162; PD-1; ribosomal protein L17; RPL17; RPL23
Gene Symbols Rpl17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL17. Recombinant protein control fragment (Product #RP-99540).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6139
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.