missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570676
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene is a member of the protein kinase C and casein kinase substrate in neurons family. The encoded protein is involved in linking the actin cytoskeleton with vesicle formation. Alternative splicing results in multiple transcript variants.
Specifications
| RPL21 | |
| Polyclonal | |
| Unconjugated | |
| Rpl21 | |
| 60S ribosomal protein L21; 8430440E03Rik; HYPT12; L21; Large ribosomal subunit protein eL21; ribosomal protein L21; ribosomal protein L21 pseudogene 16; Rpl21; RPL21P16; similar to ribosomal protein L21 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6144 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P46778 | |
| Rpl21 | |
| synthetic peptide directed towards the middle region of human RPL21 (aa 79-128). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction