missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL23A Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-22284CL1
This item is not returnable.
View return policy
Description
RPL23A Polyclonal antibody specifically detects RPL23A in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| RPL23A | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| 60S ribosomal protein L23a, FLJ27455, MDA20, melanoma differentiation-associated gene 20, ribosomal protein L23a | |
| The immunogen this antibody was made to, maps to a region between residue 106 and 156 of human Ribosomal Protein L23a using the numbering given in entry NP_000975.2 (GeneID 6147). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 6147 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction