missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590217
This item is not returnable.
View return policy
Description
Immunogen sequence: MSAHLQWMVV RNCSSFLIKR NKQTYSTEPN NLKARNSFRY NGLIHRKTVG VEPAADGKGV VVVIKRRSGQ RKPATSYVRT TINKNARATL SSIRHMIRKN; Positive Samples: HT-29, 293T, DU145, mouse kidney, mouse liver, rat spleen.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L28E family of ribosomal proteins. It is located in the cytoplasm. Variable expression of this gene in colorectal cancers compared to adjacent normal tissues has been observed, although no correlation between the level of expression and the severity of the disease has been found. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternative splicing results in multiple transcript variants encoding distinct isoforms.[provided by RefSeq, Oct 2008].
Specifications
| RPL28 | |
| Polyclonal | |
| Unconjugated | |
| Rpl28 | |
| 60S ribosomal protein L28; D7Wsu21e; hypothetical protein LOC618702; L28; Large ribosomal subunit protein eL28; ribosomal protein L28; ribosomal protein L28-like; RPL28; rpl28l; zgc:66289 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19943, 6158, 64638 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P17702, P41105, P46779 | |
| Rpl28 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RPL28 (NP_0009822). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction