missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL32 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541716
This item is not returnable.
View return policy
Description
Peptide sequence: AALRPLVKPK IVKKRTKKFI RHQSDRYVKI KRNWRKPRGI DNRVRRRFKG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 93%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L32E family of ribosomal proteins. It is located in the cytoplasm. Although some studies have mapped this gene to 3q13. 3-q21, it is believed to map to 3p25-p24. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternatively spliced transcript variants encoding the same protein have been observed for this gene.
Specifications
| RPL32 | |
| Polyclonal | |
| Unconjugated | |
| Rpl32 | |
| 60S ribosomal protein L32; AU020185; hypothetical protein LOC615556; hypothetical protein LOC677748; L32; Large ribosomal subunit protein eL32; PP9932; ribosomal protein L32; RPL32; rpL32-3A; snoRNA MBI-141; zgc:136952 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6161 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P62910 | |
| Rpl32 | |
| Synthetic peptide directed towards the N-terminal of human RPL32 (aa 2-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction