missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RPL36 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30666
This item is not returnable.
View return policy
Description
RPL36 Polyclonal antibody specifically detects RPL36 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| RPL36 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 25873 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DKFZp566B023,60S ribosomal protein L36, M, ribosomal protein L36 | |
| Between 55 and 105 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction