missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL36 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97618PCP
This item is not returnable.
View return policy
Description
RPL36 Polyclonal antibody specifically detects RPL36 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RPL36 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| DKFZp566B023,60S ribosomal protein L36, M, ribosomal protein L36 | |
| Produced in rabbits immunized with E. coli-derived Human RPL36 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 25873 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction