missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL36A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570719
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%.
RPL36A is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytosol; endoplasmic reticulum; plasma membrane; ribosome; structural constituent of ribosome; translation.
Specifications
| RPL36A | |
| Polyclonal | |
| Unconjugated | |
| RPL36A | |
| 60S ribosomal protein L36a; 60S ribosomal protein L36-A; 60S ribosomal protein L44; Cell growth-inhibiting gene 15 protein; cell migration-inducing gene 6 protein; dJ164F3.3 (ribosomal protein L44); GIG15; L36A; L39; L44L; L44-like ribosomal protein; Large ribosomal subunit protein eL36-A; Large ribosomal subunit protein eL42; MIG6; ribosomal 60S subunit protein L36A; ribosomal protein L36A; ribosomal protein L44; RPL36A; RPL39A; RPL39B; Rpl44; YL39; YM9646.06; YMR194W | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6173 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P83881 | |
| RPL36A | |
| synthetic peptide directed towards the middle region of human RPL36A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction