missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL37A Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97946IR
This item is not returnable.
View return policy
Description
RPL37A Polyclonal specifically detects RPL37A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RPL37A | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 60S ribosomal protein L37a, MGC74786, ribosomal protein L37a | |
| Produced in rabbits immunized with E. coli-derived Human RPL37A fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 6168 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction