missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL39L Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97619PE
This item is not returnable.
View return policy
Description
RPL39L Polyclonal antibody specifically detects RPL39L in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RPL39L | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| 60S ribosomal protein L39-like, L39-2, ribosomal protein L39-like, ribosomal protein L39-like 1,60S ribosomal protein L39-2, ribosomal protein L39-like protein, RPL39L1 | |
| Produced in rabbits immunized with E. coli-derived Human RPL39L fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 116832 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction