missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPL3L Polyclonal Antibody
GREENER_CHOICE

Product Code. 15623895
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15623895 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15623895 Supplier Invitrogen™ Supplier No. PA549961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total RPL3L protein.

A full-length cDNA encoding a novel ribosomal protein L3 gene was isolated and sequenced. The deduced protein sequence is 407 amino acids long and shows 77% identity to other known mammalian ribosomal protein L3 genes, which are themselves highly conserved. Southern blot analysis of human genomic DNA suggests that this novel gene is single copy. While the previously identified human ribosomal protein L3 gene has ubiquitous expression in all tissues surveyed, the novel gene described herein is strongly expressed in skeletal muscle and heart tissue, with low levels of expression in the pancreas. This novel gene, RPL3L, is located in a gene-rich region near the PKD1 and TSC2 genes on chromosome 16p13.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL3L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPL3L
Gene Accession No. Q92901
Gene Alias 1110057H16Rik; 60S ribosomal protein L3-like; Large ribosomal subunit protein uL3-like; ribosomal protein L3 like; ribosomal protein L3 pseudogene; ribosomal protein L3-like; RPL3L
Gene Symbols RPL3L
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human RPL3L (aa 211-260).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6123
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.