missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94398-0.1ml
This item is not returnable.
View return policy
Description
RPL4 Polyclonal antibody specifically detects RPL4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RPL4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| ribosomal protein L4,60S ribosomal protein L1,60S ribosomal protein L4, RPL1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human RPL4 (NP_000959.2). MACARPLISVYSEKGESSGKNVTLPAVFKAPIRPDIVNFVHTNLRKNNRQPYAVSELAGHQTSAESWGTGRAVARIPRVRGGGTHRSGQGAFGNMCRGGRMFAPTKTWRRWHRRVNTTQKRYAICSALAASALPALVMSKGHRIEEVPELPLVVEDKVEGYKKTKEAVLLLKKLKAWNDIKKVYASQRMRAGKGKMRNRRRIQRRGPCIIYNEDNGIIKAFRNIPGITLLNVSKLNILKL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6124 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction