Learn More
RPL7 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-2268C
Product Code. 18023546
  • € 639.00 / 100µL
Description
RPL7 Polyclonal specifically detects RPL7 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPL7
Polyclonal
Western Blot
humL7-1,60S ribosomal protein L7, MGC117326, ribosomal protein L7
Rabbit
Affinity Purified
RUO
Primary
Human, Mouse
IgG
Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
RPL7
The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human Ribosomal Protein 7 using the numbering given in entry NP_000962.2 (GeneID 6129).
100 μL
Immune System Diseases, Immunology
6129
Store at 4C in the dark.