Learn More
RPL7 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-2268C
Product Code. 18023546
  • € 639.00 / 100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
RPL7 Polyclonal specifically detects RPL7 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
RPL7
Polyclonal
Western Blot
humL7-1,60S ribosomal protein L7, MGC117326, ribosomal protein L7
Rabbit
Affinity Purified
RUO
Primary
Human, Mouse
IgG
Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
RPL7
The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human Ribosomal Protein 7 using the numbering given in entry NP_000962.2 (GeneID 6129).
100 μL
Immune System Diseases, Immunology
6129
Store at 4C in the dark.