Learn More
RPL9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14117-1-AP-150UL
This item is not returnable.
View return policy
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L6P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
| RPL9 | |
| Polyclonal | |
| Unconjugated | |
| RPL9 | |
| 60S ribosomal protein L9, ribosomal protein L9, RPL9, RPL9P7, RPL9P8, RPL9P9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20005, 29257, 6133 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P17077, P32969, P51410 | |
| RPL9 | |
| RPL9 Fusion Protein Ag5266 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |