missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPLP0 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97776AF750
This item is not returnable.
View return policy
Description
RPLP0 Polyclonal specifically detects RPLP0 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| RPLP0 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| 60S acidic ribosomal protein P0, 60S ribosomal protein L10E, acidic ribosomal phosphoprotein P0, L10E, MGC111226, MGC88175, P0, PRLP0, ribosomal protein, large, P0, RPP0 | |
| Produced in rabbits immunized with E. coli-derived Human RPLP0 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 6175 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction