missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPLP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15104
This item is not returnable.
View return policy
Description
RPLP2 Polyclonal specifically detects RPLP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RPLP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| RPLP2 | |
| Recombinant protein encompassing a sequence within the center region of human RPLP2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 6181 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.35mg/mL | |
| Western Blot 1:5000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| acidic ribosomal phosphoprotein P2, MGC71408,60S acidic ribosomal protein P2, P2, Renal carcinoma antigen NY-REN-44, ribosomal protein, large, P2, RPP2D11S2243E | |
| Rabbit | |
| 12 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction