missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rpmB/L28 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23514AF647
This item is not returnable.
View return policy
Description
rpmB/L28 Polyclonal antibody specifically detects rpmB/L28 in E. coli samples. It is validated for Western Blot, ELISA
Specifications
| rpmB/L28 | |
| Polyclonal | |
| Western Blot, ELISA | |
| 50S ribosomal subunit protein L28, ECK3627, Large ribosomal subunit protein bL28, rpmB | |
| Antibody was raised against an 15 amino acid peptide near the amino terminus of E. coli rpmB/L28. The immunogen is located within the first 50 amino acids of rpmB/L28. | |
| 0.1 mL | |
| Primary | |
| E. coli | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 946941 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction