missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPN1 Monoclonal Antibody (OTI5B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503655S
This item is not returnable.
View return policy
Description
RPN1 Monoclonal antibody specifically detects RPN1 in Canine, Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| RPN1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| E2RQ08, P04843 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 476516, 6184 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5B1 | |
| Unconjugated | |
| RPN1 | |
| AU018702; D6Wsu137e; Dolichyl-diphosphooligosaccharide--protein; dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1; oligosaccharyltransfe; oligosaccharyltransferase 1 homolog; oligosaccharyltransferase complex subunit (non-catalytic); OST1; QccE-21639; QtsA-18349; RBPH1; RIBI; ribophorin I; Ribophorin-1; RPN1; Rpn-1; RPN-I; unnamed protein product | |
| Full length human recombinant protein of human RPN1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction