Learn More
RPN1 Mouse anti-Canine, Human, Clone: OTI2G1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand:  Origene Technologies CF503654
Encompass_Preferred
Product Code. 17603301
  • POA / 100µg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Ribophorins 1 and 2 are abundant and highly conserved glycoproteins residing in the endoplasic reticulum, that participate in ribosome binding. Mammalian oligosaccharyltransferase activity is associated with a protein complex composed of RPN1, RPN2, and an oligosaccharyltransferase protein. RPN1 is a component of the proteasome base. The ubiquitin-like (UBL) domain of recombit Rad23 interacts with proteasomes through the leucine-rich repeat domain of RPN1. The RPN1 gene maps to chromosome 3 in somatic cell hybrids, and the RPN2 gene maps to chromosome 20 by in situ hybridization.
Specifications
Specifications
RPN1
Monoclonal
Unconjugated
RPN1
OST1, RBPH1
Mouse
Affinity Chromatography
RUO
476516, 6184
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2G1
PBS with 8% trehalose and no preservative; pH 7.3
E2RQ08, P04843
RPN1
Full length human recombit protein of human RPN1 produced in HEK293T cell.
100 μg
Primary
Canine, Human