missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPP21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107017
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 93%.
RPP21 is a protein subunit of nuclear ribonuclease P, which processes the 5-prime leader sequence of precursor tRNAs.
Specifications
| RPP21 | |
| Polyclonal | |
| Unconjugated | |
| Rpp21 | |
| 0610037N01Rik; C6orf135; Cat60; Flj22638; ribonuclease P 21 subunit; ribonuclease P 21kDa subunit; ribonuclease P protein subunit p21; Ribonuclease P/MRP 21 kDa subunit; ribonuclease P/MRP 21 subunit; ribonuclease P/MRP 21kDa subunit; ribonuclease P/MRP subunit p21; ribonucleoprotein V; RNaseP protein p21; Rpp21 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9H633 | |
| Rpp21 | |
| Synthetic peptide located within the following region: LVPGLTCTQR QRRCRGQRWT VQTCLTCQRS QRFLNDPGHL LWGDRPEAQL. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction