missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPP25 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94328-0.1ml
This item is not returnable.
View return policy
Description
RPP25 Polyclonal antibody specifically detects RPP25 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RPP25 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| FLJ20374, ribonuclease P 25kDa subunit, ribonuclease P protein subunit p25, ribonuclease P/MRP 25kDa subunit, RNase P protein subunit p25 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of human RPP25 (NP_060263.2). MENFRKVRSEEAPAGCGAEGGGPGSGPFADLAPGAVHMRVKEGSKIRNLMAFATASMAQPATRAIVFSGCGRATTKTVTCAEILKRRLAGLHQVTRLRYRSVREVWQSLPPGPTQGQTPGEPAASLSVLKNVPGLAILLSKDALDPRQPGYQPPNPHPGPSSPPAAPASKRSLGEPAAGEGSAKRSQPEPGVADEDQTA | |
| 0.1 mL | |
| Endocrinology | |
| 54913 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction