missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPRD1B Antibody (01), DyLight 594 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06202DL594
This item is not returnable.
View return policy
Description
RPRD1B Monoclonal antibody specifically detects RPRD1B in Human samples. It is validated for Western Blot
Specifications
| RPRD1B | |
| Monoclonal | |
| DyLight 594 | |
| Cell cycle-related and expression-elevated protein in tumor, cell-cycle related and expression-elevated protein in tumor, chromosome 20 open reading frame 77, CREPTC20orf77, dJ1057B20.2, DKFZp434P0735, FLJ44520, NET60, regulation of nuclear pre-mRNA domain containing 1B, regulation of nuclear pre-mRNA domain-containing protein 1B | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RPRD1B (AAH33629.1; Met1-Asp326). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 01 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 58490 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction